Order Entry
Austria
ContactUsLinkComponent
N,N-Dimethylacetamide ≥99.5% (by GC), BAKER ANALYZED®, J.T.Baker®
Health
Irritant
N,N-Dimethylacetamide ≥99.5% (by GC), BAKER ANALYZED®, J.T.Baker®
Catalog # J372-07
Supplier:  Avantor
CAS Number:  
undefined
Health
Irritant
N,N-Dimethylacetamide ≥99.5% (by GC), BAKER ANALYZED®, J.T.Baker®
Catalog # J372-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (CH₃CON(CH₃)₂) (by GC):
    ≥99.5 %
  • Density (g/mL) at 25°C:
    0.931 - 0.936
  • Residue after Ignition:
    ≤0.005 %
  • pH of 20% Solution at 25°C:
    4.0 - 7.0
  • Water (H₂O):
    ≤0.05 %

Specification Test Results

About this item

 :
 :87.12 g/mol
Merck Index:13,03254
Boiling Pt:166.1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.937 g/cm³ (20 °C)
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
UN:2810
ADR:6.1,III
 :GHS08
 :GHS07
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686