Orders LinkContactUsLinkComponent
VWR®, Camera Adapters
Supplier:   VWR Collection
VWR®, Camera Adapters
Supplier:  VWR Collection
Certificates
VWR®, Camera Adapters
5 Options Available Below
Product Details & Documents
These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
Product Family Options
Product Information
  • Availability
    Price
  • 630-0867
    C-mount adapter 0,4×
    C-mount adapter 0,4×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,60 €
    42,60 €
    1 * 1 ST
       42,60 €
  • 630-0865
    C-mount adapter 0,5×
    C-mount adapter 0,5×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,00 €
    62,00 €
    1 * 1 ST
       62,00 €
  • 630-2687
    C-mount adapter 0,45× for INV100/100-FL
    C-mount adapter 0,45× for INV100/100-FL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,60 €
    86,60 €
    1 * 1 ST
       86,60 €
  • 630-0866
    C-mount adapter 1,0×
    C-mount adapter 1,0×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,50 €
    48,50 €
    1 * 1 ST
       48,50 €
  • 630-0868
    C-mount adapter 1,0×
    C-mount adapter 1,0×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,90 €
    16,90 €
    1 * 1 ST
       16,90 €