Disposable earplugs, uvex com4-fit
Supplier: UVEX
Certificates
About this item
Thanks to their compact design, the com4-fit earplugs are ideal for wearers with narrow ear canals.
- Ergonomic shape adapts to the natural shape of the ear canal
- Combines optimal comfort with the highest level of protection
- x-grip technology reduces expansion pressure and therefore increases wearer comfort, even when worn for extended periods
- Easy insertion and removal
- For small to medium ear canals
- High visibility colour for easy visual recognition
SNR: 33 dB
Certifications: EN 352-2:2002
Frequently Bought Together