Racks for micro tubes, floating , SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Molded of steam autoclavable polypropylene, these racks fit 400 ml or 1000 ml beakers.
- For 1,5 ml micro tubes
- Tubes remain completely immersed in a beaker while tops rest above the rack
- Unload by lightly pressing down against the bench until the legs touch, releasing the tubes
Frequently Bought Together