Orders LinkContactUsLinkComponent
Threaded couplings
Threaded couplings
Supplier:   Bohlender
Threaded couplings
Supplier:  Bohlender
Certificates
Threaded couplings
Threaded couplings
10 Options Available Below
Product Details & Documents
Connector, screw thread coupling, made of PTFE. Two screw caps made of PPS or PBTP with GL thread and a PTFE/FPM gasket. The product is only exposed to PTFE.
  • Applicable for connecting two GL threads of different sizes
  • Temperature resisitance from −200 to +250 ºC
  • Very good chemical resistance
  • FDA conformity
Product Family Options
With PPS cap
Product Information
  • Thread
    Availability
    Price
  • GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,40 €
    30,40 €
    1 * 1 ST
       30,40 €
  • GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,40 €
    43,40 €
    1 * 1 ST
       43,40 €
  • GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,00 €
    39,00 €
    1 * 1 ST
       39,00 €
  • GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,20 €
    80,20 €
    1 * 1 ST
       80,20 €
  • GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,10 €
    65,10 €
    1 * 1 ST
       65,10 €
With PBTB cap
Product Information
  • Thread
    Availability
    Price
  • GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,60 €
    22,60 €
    1 * 1 ST
       22,60 €
  • GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,70 €
    50,70 €
    1 * 1 ST
       50,70 €
  • GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,20 €
    23,20 €
    1 * 1 ST
       23,20 €
  • GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,70 €
    34,70 €
    1 * 1 ST
       34,70 €
  • GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,60 €
    37,60 €
    1 * 1 ST
       37,60 €