ABsolute™ qPCR Capillary Mixes
ABsolute™ qPCR Capillary Mixes contain an optimised reaction buffer specifically designed to provide the highest possible sensitivity, specificity and consistent end-point values in real time instruments that use glass capillaries, such as the Roche LightCycler®. The 2X mixes contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes.
- Contains hot start enzyme Thermo-Start® DNA polymerase for increased specificity and optimised buffer for maximum sensitivity
- Excellent reproducibility, with all batches undergoing extensive quality control to ensure batch-to-batch and assay-to-assay consistency
- Optimised buffer specifically designed for real-time instruments that use glass capillaries
- Application:qPCR probe
Frequently Bought Together