Orders LinkContactUsLinkComponent
DNA polymerases
DNA polymerases
DNA polymerases
Certificates
9 Options Available Below
Product Details & Documents
KOD HiFi DNA Polymerase is a unique proofreading enzyme, isolated from the extreme thermophile Thermococcus kodakaraensis KOD1, which possesses superior processivity and fidelity that enables faster and more accurate PCR amplification than can be achieved with conventional enzymes.
KOD HiFi DNA Polymerase is also available in a hot start version for high specificity and increased read length, and as a blend (KOD XL DNA Polymerase) recommended for very long templates.
NovaTaq™ DNA Polymerase is a high-purity, recombinant enzyme suitable for any application requiring premium quality Taq DNA polymerase.
For increased specificity and convenience with standard PCR, NovaTaq Hot Start DNA Polymerase is a chemically modified form of the enzyme that becomes active when heated at 95 °C for 7 to 10 minutes.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 71003-3
    NovaTaq DNA polymerase
    NovaTaq DNA polymerase
    100 EU
    Log in for availability
    Log in for availability
    1 * 100 EU
    1 * 100 EU - 75,80 €
    75,80 €
    1 * 100 EU
       75,80 €
  • 71003-4
    NovaTaq DNA polymerase
    NovaTaq DNA polymerase
    500 EU
    Log in for availability
    Log in for availability
    1 * 500 EU
    1 * 500 EU - 312,00 €
    312,00 €
    1 * 500 EU
       312,00 €
  • 71003-5
    NovaTaq DNA polymerase
    NovaTaq DNA polymerase
    2.500 EU
    Log in for availability
    Log in for availability
    1 * 2.500 EU
    1 * 2.500 EU - 1 280,00 €
    1 280,00 €
    1 * 2.500 EU
       1 280,00 €
  • 71087-3
    KOD XL DNA polymerase
    KOD XL DNA polymerase
    250 EU
    Log in for availability
    Log in for availability
    1 * 250 EU
    1 * 250 EU - 600,00 €
    600,00 €
    1 * 250 EU
       600,00 €
  • 71091-3
    NovaTaq Hot Start DNA polymerase
    NovaTaq Hot Start DNA polymerase
    250 EU
    Log in for availability
    Log in for availability
    1 * 250 EU
    1 * 250 EU - 473,00 €
    473,00 €
    1 * 250 EU
       473,00 €
  • 71091-4
    NovaTaq Hot Start DNA polymerase
    NovaTaq Hot Start DNA polymerase
    1.250 EU
    Log in for availability
    Log in for availability
    1 * 1.250 EU
    1 * 1.250 EU - 2 200,00 €
    2 200,00 €
    1 * 1.250 EU
       2 200,00 €
  • 71086-4
    KOD Hot Start DNA polymerase
    KOD Hot Start DNA polymerase
    1.000 EU
    Log in for availability
    Log in for availability
    1 * 1.000 EU
    1 * 1.000 EU - 1 560,00 €
    1 560,00 €
    1 * 1.000 EU
       1 560,00 €
  • 71085-3
    KOD HiFi DNA polymerase
    KOD HiFi DNA polymerase
    250 EU
    Log in for availability
    Log in for availability
    1 * 250 EU
    1 * 250 EU - 507,00 €
    507,00 €
    1 * 250 EU
       507,00 €
  • 71086-5
    KOD Hot Start DNA polymerase
    KOD Hot Start DNA polymerase
    20 EU
    Log in for availability
    Log in for availability
    1 * 20 EU
    1 * 20 EU - 106,00 €
    106,00 €
    1 * 20 EU
       106,00 €