ContactUsLinkComponent
Tris-Borate-EDTA (TBE) buffer, 10X, Molecular biology grade
Tris-Borate-EDTA (TBE) buffer, 10X, Molecular biology grade
Catalog # A3945.1000
Supplier:  PanReac AppliChem
CAS Number:  
Tris-Borate-EDTA (TBE) buffer, 10X, Molecular biology grade
Catalog # A3945.1000
Supplier:  PanReac AppliChem
CAS Number:  
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Tris-Borate-EDTA (TBE) buffer, 10X, Molecular biology grade
Specifications
About this item
10X concentrated solution of 0,89 M Tris, 0,89 M boric acid and 0,02 M EDTA.Na₂, pH 8,3 ±0,2.
DNases, RNases, and proteases not detectable.