Order Entry
Austria
ContactUsLinkComponent
VWR® Phosphate Buffered Saline (PBS), Low Phosphate
Catalog # 444057Y
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Phosphate Buffered Saline (PBS), Low Phosphate
Catalog # 444057Y
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Description:
    PBS buffer (phosphate buffered saline) with low phosphate

Specifications

About this item

Irradiated solution for use in washing cells.

  • KCl 0,2 g/L
  • NaCl 7,01 g/L
  • Na₂HPO₄, 12H₂0 1,28 g/L
  • NaH₂PO₄, 2H₂O 0,44g/L