VWR® Ready to Use Media Plates (90 mm), Irradiated
Supplier: VWR Chemicals
Certificates
About this item
Available as 90 mm plates acc. to standards. TLHTh (Tween®, lecithin, histidine, thiosulphate) are used as disinfectant neutralisers. Bags are available double- or triple-wrapped. One pack per bag displays the irradiation indicator.
- Triple-wrapping ensures that the package itself doesn't contaminate the environment, as the first wrapper is removed just before entering a clean area
- Available with certificate of irradiation/dosimetry
- VHP resistance studies available for all triple-wrapped plates
Storage: 15 to 25 °C for plates 280882TI, 280882ZI, 281112TI and 281112ZI; 2 to 14 °C for all the other plates.
Packaging: The plates are packed 10 each in a plastic shrink-wrapped bag. Box contains 2 bags with a total of 20 or 6×10 plates.
Related Information
Certificates
Search by Lot Number110070PY_ROSE BENGAL CHLOR.PENASE 90MM DOUBLE-WR._ISO 11133_rev1 110774ZI_PCA PETRI 90MM DOUBLE-WRAP_ISO 11133_rev1 110774ZIMP_PCA PETRI 90MM DOUBLE-WRAP_ISO 11133_rev1 110802ZI_R2A PETRI 90MM DOUBLE-WRAP_ISO 11133_rev1 110802ZIMP_R2A PETRI 90MM DOUBLE-WRAP_ISO 11133_rev1
More
SDS_EN_UK_VWRC110774ZI.pdf SDS_EN_UK_VWRC110884ZI.pdf SDS_EN_UK_VWRC111114TI.pdf SDS_EN_UK_VWRC111114ZI.pdf SDS_EN_UK_VWRC112230CA.pdf
SDS
More
spec_131114TI spec_VWRC110774ZI spec_VWRC110802ZI spec_VWRC110884CY spec_VWRC110884TI
Product specification
More
Frequently Bought Together