Order Entry
Austria
ContactUsLinkComponent
Ready-to-use media, Merckoplate®
Ready-to-use media, Merckoplate®
Catalog # 1.13405.0001
Supplier:  Merck
CAS Number:  
undefined
Ready-to-use media, Merckoplate®
Catalog # 1.13405.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    480
  • Standards:
    -
  • Description:
    Mueller-Hinton agar
  • Filling:
    20 ml

Specifications

About this item

The Merckoplate® product range includes a full line of pre-poured media for a wide range of different applications, such as enumeration, isolation, detection and cultivation of bacteria as well as yeast and moulds. In order to reduce waste, bulk packs of 480 plates are available for customers with high plate throughput. The majority of Merckoplate® products can be stored at a temperature below 15 °C, although, in order to minimise condensation, the optimal storage temperature is 12 to 15 °C (Do not freeze!).

  • Ready to use media in 90 mm Petri dishes
  • Item number and the expiry date are imprinted on every plate for safe and easy product identification and shelf life monitoring
Packaging: 4 or 96 sets of 5 shrink-wrapped plates