VWR® Contact Plates for Surface and Air Control, Irradiated
Supplier: VWR Chemicals
Certificates
About this item
Contact plates are ideal for monitoring microbiological contamination of surfaces and air (using an air sampler) inside cleanrooms, isolators, RABS (Restricted Access Barrier), the food industry and hospitals. Triple-wrapping ensures that the package itself does not contaminate the environment, as the first wrapper is removed just before entering a clean area. Plate packaging containing PCA is not irradiated.
- Plates packed in special triple-wrapped packaging with irradiation indicator
- Can use PET, transparent, reusable container to transport plates to the laboratory after sampling
- Special packaging to protect the product during transport and storage
- Available with TLHTh (Tween®, lecithin, histidine, thiosulphate) disinfectant neutralisers
- Available with certificate of irradiation/dosimetry
- VHP resistance studies available for all triple-wrapped plates
Packaging: Contains 8×10 triple-wrapped packs with a total of 80 plates. 8 PET, transparent, reusable containers are included.
Related Information
Certificates
Search by Lot Number140070ZI_RBC AGAR CONTACT 55MM TRIPLE-WRAP_ISO 11133_rev1 140774TI_PCA TLHTH CONTACT 55MM TRIPLE-WRAP_ISO 11133_rev1 140774ZI_PCA CONTACT 55MM TRIPLE-WRAP_ISO 11133_rev1 140884CY_SABOURAUD CHLORAMPHENICOL CONTACT_ISO 11133_rev1 140884TI_SAB 4%DEX AGAR CONTACT TLHTH TRIPLE-WR_ISO 11133_rev1
More
BSE TSE 141114ZI TSA contact triple wrap
BSE/TSE Certificates
SDS_EN_CH_VWRC140774ZI.pdf SDS_EN_DE_VWRC140774ZI.pdf SDS_EN_UK_VWRC140774TI.pdf SDS_EN_UK_VWRC140774ZI.pdf SDS_EN_UK_VWRC140884ZI.pdf
SDS
More
spec_141114ZI spec_VWRC140070ZI spec_VWRC140774TI spec_VWRC140774ZI spec_VWRC140884CY
Product specification
More
Frequently Bought Together