Order Entry
Austria
ContactUsLinkComponent
Standard solutions for analytical quality assurance, Supelco®
Standard solutions for analytical quality assurance, Supelco®
Catalog # 1.09017.0100
Supplier:  Merck
CAS Number:  
Standard solutions for analytical quality assurance, Supelco®
Catalog # 1.09017.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 (mg/l)
  • Pk:
    100 mL
  • Description:
    TOC acc. to EN 1484 H3
  • M-Clarity quality level:
    MQ100

Specifications

About this item

The standard solutions offered below can be adjusted to a final concentration for the measurement range of a test system by dilution. In addition, these standards can be used to prepare spiking solutions for the standard addition procedure. They provide an economic and simple method of analytical quality control.

All CertiPUR® standard solutions can be traced back to NIST.