Orders LinkContactUsLinkComponent
Crack sets for sample preparation, Spectroquant®, Supelco®
 :   Merck
Crack sets for sample preparation, Spectroquant®, Supelco®
 :  Merck
Crack sets for sample preparation, Spectroquant®, Supelco®
Complete sets for digestion in a thermoreactor. The subsequent determination can be carried out in Spectroquant® test cells or reagent test sets. Crack Sets are easy to use and make sample preparation straightforward. For the preparation of dirty, coloured or samples with high levels of suspended material prior to using the appropriate Spectroquant® kit.
  • Contains all reagents required for selected tests
  • Easy to use - with step-by-step instructions
  • Catalog No:
  • 1.14687.0001
  • 1.14688.0001
  • 1.14963.0001
  • Model:
  • Crack Set 10
  • Crack Set 10C
  • Crack Set 20
  • Method:
  • Peroxodisulfate/acidic
  • Peroxodisulfate/acidic
  • Korolef
  • Parameters:
  • Pb, Cd, Cr, Fe, Cu, Ni, Zn and P(total)
  • Pb, Cd, Cr, Fe, Cu, Ni, Zn and P(total)
  • Total nitrogen
  • Contents:
  • Digestion reagent, acid, neutralising agent for pH adjustment.
  • Digestion reagent in round cells, acid, neutralising agent for pH adjustment.
  • Digestion reagent, base.
  • No. of digestions:
  • 100
  • 25
  • 90
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
Product Family Options
  • Description
    Pk
  • Crack Set 10 Spectroquant®
    100 Tests
    1 * 100 Tests
    1 * 100 Tests - 191,00 €
    191,00 €
    1 * 100 Tests
       191,00 €
    Specifications
    Cat. No.
    1.14687.0001
    Model
    Crack Set 10
    Method
    Peroxodisulfate/acidic
    Parameters
    Pb, Cd, Cr, Fe, Cu, Ni, Zn and P(total)
    Contents
    Digestion reagent, acid, neutralising agent for pH adjustment.
    No. of digestions
    100
    M-Clarity quality level
    MQ100
  • Crack Set 10C Spectroquant®
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 181,00 €
    181,00 €
    1 * 25 Tests
       181,00 €
    Specifications
    Cat. No.
    1.14688.0001
    Model
    Crack Set 10C
    Method
    Peroxodisulfate/acidic
    Parameters
    Pb, Cd, Cr, Fe, Cu, Ni, Zn and P(total)
    Contents
    Digestion reagent in round cells, acid, neutralising agent for pH adjustment.
    No. of digestions
    25
    M-Clarity quality level
    MQ100
  • Crack Set 20 Spectroquant®
    90 Tests
    1 * 90 Tests
    1 * 90 Tests - 177,00 €
    177,00 €
    1 * 90 Tests
       177,00 €
    Specifications
    Cat. No.
    1.14963.0001
    Model
    Crack Set 20
    Method
    Korolef
    Parameters
    Total nitrogen
    Contents
    Digestion reagent, base.
    No. of digestions
    90
    M-Clarity quality level
    MQ100
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.