Photometric test kit for iron
705-0117
: Hach
:
- Chemical method:Ferrozine
- Pk:50
- Description:Iron, liquid reagents* in pillows
- Range:0,009 - 1,400 mg/l
For determination of trace levels of iron by the Ferrozine method.
For determination of trace levels of iron by the Ferrozine method.