Accessories for titrators
Supplier: Mettler - Toledo
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Accessories for Mettler balances
Supplier: Mettler - Toledo
Accessories for Magnetic Hotplate Stirrers
Supplier: IKA
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
pH electrodes, SenTix® PLUS
Supplier: WTW
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Accessories for titrators
Supplier: Mettler - Toledo
Plastic cuvette for LICO
Supplier: Hach