Order Entry
Austria
ContactUsLinkComponent
pH/mV/°C meters, handheld, pH 3110 / 3310
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier:  WTW
Certificates
undefined
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier:  WTW

Specifications

  • Catalog No:
  • 662-1111
  • 662-0999
  • 662-0503
  • 662-0505
  • Model:
  • pH 3310
  • pH 3310
  • pH 3110
  • pH 3110
  • pH range:
  • −2,0...+19,9 / −2,00...+19,99 / −2,000...+19,999
  • −2,0...+19,9 / −2,00...+19,99 / −2,000...+19,999
  • −2,0...+19,9 / −2,00...+19,99 / −2,000...+19,999
  • −2,0...+19,9 / −2,00...+19,99 / −2,000...+19,999
  • pH accuracy:
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • pH calibration:
  • 1, 2, 3, 4 or 5 points with 24 sets of memorised buffers
  • 1, 2, 3, 4 or 5 points with 24 sets of memorised buffers
  • 1, 2 or 3 points with WTW or DIN buffers
  • 1, 2 or 3 points with WTW or DIN buffers
  • mV range:
  • ±1200,0 / ±2500
  • ±1200,0 / ±2500
  • ±1200,0 / ±2000
  • ±1200,0 / ±2000
  • mV accuracy:
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • Temperature range:
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • −5,0...+105,0 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Display:
  • Backlit graphic LCD
  • Backlit graphic LCD
  • LCD
  • LCD
  • Power supply:
  • 4× 1,5 V AA or 4× 1,2 V NiMH
  • 4× 1,5 V AA or 4× 1,2 V NiMH
  • 4× 1,5 V AA or 4× 1,2 V NiMH
  • 4× 1,5 V AA or 4× 1,2 V NiMH
  • Compliance:
  • IP 66/67, GLP
  • IP 66/67, GLP
  • IP 66/67
  • IP 66/67
  • Data logging - log function:
  • Manual / time-controlled
  • Manual / time-controlled
  • -
  • -
  • Data logging capacity:
  • 200 manual / 5000 automatic
  • 200 manual / 5000 automatic
  • -
  • -
  • Memory:
  • Last 5
  • Last 5
  • Current calibration
  • Current calibration
  • Connections:
  • DIN / 4 mm banana
  • DIN / 4 mm banana
  • DIN / 4 mm banana
  • DIN / 4 mm banana
  • Outputs:
  • USB
  • USB
  • -
  • -
  • Weight:
  • 0,6 kg
  • 0,6 kg
  • 0,6 kg
  • 0,6 kg

Specifications

Electrodes

Product Information

Frequently Bought Together