Dimroth condensers for Soxleth extractors
DURAN® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Dimroth condensers for extractors, for condensing vapours. Water connection with two glass olives.
Frequently Bought Together