ContactUsLinkComponent
TLC spray cabinets
TLC spray cabinets
  CAMA022.6230UK
 :  CAMAG
undefined
TLC spray cabinets
  CAMA022.6230UK
 :  CAMAG
  • Description:
    TLC spray cabinet II, UK-plug, 470×490×490 mm, with blower and 1,5 m flexible exhaust hose (127 mm Ø)
  • Seq:
    02
  • Pk:
    1
The TLC spray cabinet is designed for the complete removal of excessive spray mist while spraying a TLC plate with reagent.
  • Made of PVC
  • The blower, a radial fan driven by a motor outside of the fume duct, produces an airflow of 3,7 m³/min
  • Removable built in tray, easy for cleaning
There is no deflection of the spray jet before it reaches the plate, an effect often encountered in a normal laboratory fume hood. Particles rebounding from the plate are completely removed. This spray cabinet is also useful for drying plates after development, with or without the assistance of a hair dryer.