Order Entry
Austria
Orders LinkContactUsLinkComponent
 

HPTLC basic kits

HPTLC basic kit, including UV cabinet, for qualitative and quantitative TLC analysis using 200×100 mm HPTLC layers
  CAMA022.0227
 :   CAMAG
HPTLC basic kits
  CAMA022.0227
 :  CAMAG
HPTLC basic kits

HPTLC basic kit, including UV cabinet, for qualitative and quantitative TLC analysis using 200×100 mm HPTLC layers

 
 
   
1 * 1 KIT
 

HPTLC basic kit includes:
Nanomat 4, without capillary pipettes and holder
Capillary dispenser complete with universal capillary holder, one dispenser magazine for 1 µl capillaries and one box of 5×100 capillary pipettes of 1,0 µl
Dispenser magazine for 0,5 µl capillaries
Disposable capillary pipettes 0,5 µl, on belts, 5×100 pack
Two twin trough chambers for 100×100 or 200×100 mm plates, with stainless steel lid
Horizontal developing chamber for 100×100 or 200×100 mm plates
smartALERT, solvent front monitor
Saturation pads, pack of 100
TLC/HPTLC sprayer
Precoated plates, Merck HPTLC silica gel 60 F254, 100×100 or 200×100 mm, pack of 25
Test dye mixture III, 100 ml
UV cabinet 3 consisting of UV lamp dual wavelength, 254/366 nm, 8 W each

Dispenser magazine for 2,0 µl capillaries

Dispenser magazine for 5,0 µl capillaries

Disposable capillary pipettes 2,0 µl, on belts, 5×100 pack

Disposable capillary pipettes 5,0 µl, on belts, 5×100 pack

 
  • Description:
    HPTLC basic kit, including UV cabinet, for qualitative and quantitative TLC analysis using 200×100 mm HPTLC layers
  • Seq:
    01