0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Closures
Chromatography Vial Closures
Caps with septa for screw top vials, 15×45 mm, WHEATON®
Caps with septa for screw top vials, 15×45 mm, WHEATON®
WHEA242247
:
DWK Life Sciences
Caps with septa for screw top vials, 15×45 mm, WHEATON®
WHEA242247
:
DWK Life Sciences
Type:
Preassembled open top caps and septa
Description:
Natural, 13-425, with 0,25 mm septa
Pk:
1.000