Order Entry
Austria
Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type, WHEATON®

Sedimentation cones, Imhoff type, WHEATON®

with screw cap and removal plug
Catalog # WHEAW990800
Supplier:   DWK Life Sciences
Sedimentation cones, Imhoff type, WHEATON®
Catalog # WHEAW990800
Supplier:  DWK Life Sciences
Sedimentation cones, Imhoff type, WHEATON®
Sedimentation cones, Imhoff type, WHEATON®

with screw cap and removal plug

Price
297,00 €
Log in for availability
1 * 4 ST
Product Details & Documents

Made of SAN.

For Volumetric determining the amount of settleable solids.

Graduation: 0 to 1 ml/0,1 ml; 1 to 10 ml/0,5 ml; 10 to 40 ml/1 ml; 40 to 100 ml/2 ml.

Specifications
  • Type:
    with screw cap and removal plug
  • Cat. No.:
    WHEAW990800
  • Graduation:
    0 - 1 ml
    1 - 10 ml
    10 - 40 ml
    40 - 100 ml
  • Tolerance:
    0,1 (±ml)
    0,5 (±ml)
    1 (±ml)
    2 (±ml)
  • Division:
    0,1 ml
    0,5 ml
    1 ml
    2 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.