Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Supplier:   WTW
Conductivity standards
Supplier:  WTW
Certificates
Conductivity standards
Conductivity standards
4 Options Available Below
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 663-1891
    E-Set Trace calibration set, 6 bottles with 50 ml calibration and control standard each, 0,01 mol/l KCl, 1413 µS/cm at 25 °C, accuracy ± 0.5 %, traceable to PTB/NIST
    E-Set Trace calibration set, 6 bottles with 50 ml calibration and control standard each, 0,01 mol/l KCl, 1413 µS/cm at 25 °C, accuracy ± 0.5 %, traceable to PTB/NIST
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 150,00 €
    150,00 €
    1 * 1 SET
       150,00 €
  • WTWA300578_U
    Conductivity calibration solution 100 µS/cm, shelf life 2 years, NIST traceable, accuracy ±3%
    Conductivity calibration solution 100 µS/cm, shelf life 2 years, NIST traceable, accuracy ±3%
    300 mL
    Log in for availability
    Log in for availability
    1 * 300 mL
    1 * 300 mL - 170,00 €
    170,00 €
    1 * 300 mL
       170,00 €
  • WTWA300580_U
    Conductivity calibration solution 5 µS/cm, shelf life 1 year, NIST traceable, accuracy ±2%
    Conductivity calibration solution 5 µS/cm, shelf life 1 year, NIST traceable, accuracy ±2%
    300 mL
    Log in for availability
    Log in for availability
    1 * 300 mL
    1 * 300 mL - 322,00 €
    322,00 €
    1 * 300 mL
       322,00 €
  • 663-8119
    Test set 6R/SET/LAB1 for conductivity meters
    Test set 6R/SET/LAB1 for conductivity meters
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 638,00 €
    638,00 €
    1 * 1 ST
       638,00 €