Orders LinkContactUsLinkComponent
Protective rubber case for instruments
Protective rubber case for instruments
Protective rubber case for instruments, CRS/6 for Comark instruments N5000 N8000 and N9000
Catalog # 620-1476
Supplier:   COMARK
Protective rubber case for instruments
Catalog # 620-1476
Supplier:  COMARK
Protective rubber case for instruments
Protective rubber case for instruments
Protective rubber case for instruments, CRS/6 for Comark instruments N5000 N8000 and N9000
Price
79,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
The CRS/6 protective rubber case can be used with Comark instruments N5000 N8000 and N9000.
  • Built-in probe rest
  • Carry strap
Specifications
  • Description:
    Protective rubber case for instruments, CRS/6 for Comark instruments N5000 N8000 and N9000