Orders LinkContactUsLinkComponent
Sample cuvettes
Sample cuvettes
Supplier:   Hach
Sample cuvettes
Supplier:  Hach
Certificates
Sample cuvettes
20 Options Available Below
Product Details & Documents
Sample cuvettes for photometric measurements.
  • Available with or without lid
  • Available reusable (made from glass) or for single use (made from plastics)
Product Family Options
Square base
Product Information
  • Volume
    Path length
    Window material
    Availability
    Price
  • HACH2629500
    Sample cuvettes
    Sample cuvettes
    1,5 ml
    10 mm
    PS
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 114,00 €
    114,00 €
    1 * 100 ST
       114,00 €
  • 705-0127
    Sample cuvettes, with lid, disposable
    Sample cuvettes, with lid, disposable
    25 ml
    1" / 25 mm
    PS
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 237,00 €
    237,00 €
    1 * 12 ST
       237,00 €
  • 705-0084
    Sample cuvettes, with lid, matched pair
    Sample cuvettes, with lid, matched pair
    3,5 ml
    10 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 327,00 €
    327,00 €
    1 * 2 ST
       327,00 €
  • HACH2624410
    Sample cuvette, with lid
    Sample cuvette, with lid
    3,5 ml
    10 mm
    Quartz Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 349,00 €
    349,00 €
    1 * 1 ST
       349,00 €
  • 705-0228
    Sample cuvettes, with lid, matched pair
    Sample cuvettes, with lid, matched pair
    3,5 ml
    10 mm
    Quartz Glass
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 864,00 €
    864,00 €
    1 * 2 ST
       864,00 €
  • HACHLZP045
    Sample cuvettes
    Sample cuvettes
    3,5 ml
    10 mm
    Optical glass
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 141,00 €
    141,00 €
    1 * 3 ST
       141,00 €
  • 705-0762
    Sample cuvettes, matched pair, 10 ml marked
    Sample cuvettes, matched pair, 10 ml marked
    10 ml
    1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 331,00 €
    331,00 €
    1 * 2 ST
       331,00 €
  • HACH2495408
    Sample cuvettes, matched set, 10 ml marked
    Sample cuvettes, matched set, 10 ml marked
    10 ml
    1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 834,00 €
    834,00 €
    1 * 1 SET
       834,00 €
  • HACH2612602
    Sample cuvettes, with lid, matched pair, 10/25 ml marked
    Sample cuvettes, with lid, matched pair, 10/25 ml marked
    25 ml
    1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 429,00 €
    429,00 €
    1 * 2 ST
       429,00 €
  • 705-0171
    Sample cuvettes, matched pair, 10/25 ml marked
    Sample cuvettes, matched pair, 10/25 ml marked
    25 ml
    1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 391,00 €
    391,00 €
    1 * 2 ST
       391,00 €
  • HACHLZP334
    Flow through cuvette, micro, CH = 8,5 mm
    Flow through cuvette, micro, CH = 8,5 mm
    80 µl
    10 mm
    Optical glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 912,00 €
    912,00 €
    1 * 1 ST
       912,00 €
Rectangular base
Product Information
  • Volume
    Path length
    Window material
    Availability
    Price
  • HACHLZP341
    Sample cuvettes, with lid, semi-micro
    Sample cuvettes, with lid, semi-micro
    7 ml
    50 mm
    PMMA
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 60,30 €
    60,30 €
    1 * 10 ST
       60,30 €
  • 705-0166
    Sample cuvette, with lid
    Sample cuvette, with lid
    17,5 ml
    50 mm
    Quartz Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 699,00 €
    699,00 €
    1 * 1 ST
       699,00 €
  • 705-0575
    Sample cuvette, with lid
    Sample cuvette, with lid
    17,5 ml
    50 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 286,00 €
    286,00 €
    1 * 1 ST
       286,00 €
  • HACHLZP167
    Sample cuvette
    Sample cuvette
    20 ml
    50 mm
    Optical glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 178,00 €
    178,00 €
    1 * 1 ST
       178,00 €
  • 705-0165
    Sample cuvette, with lid
    Sample cuvette, with lid
    35 ml
    100 mm
    Quartz Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 836,00 €
    836,00 €
    1 * 1 ST
       836,00 €
  • HACH2629201
    Sample cuvette, with lid
    Sample cuvette, with lid
    35 ml
    100 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 538,00 €
    538,00 €
    1 * 1 ST
       538,00 €
Round base
Product Information
  • Volume
    Path length
    Window material
    Availability
    Price
  • HACHLCW906
    Sample cuvettes, with rubber stopper, barcoded
    Sample cuvettes, with rubber stopper, barcoded
    7 ml
    13 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 72,70 €
    72,70 €
    1 * 25 ST
       72,70 €
  • 705-0749
    Sample cuvette, with lid, 10 ml marked
    Sample cuvette, with lid, 10 ml marked
    10 ml
    1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,00 €
    14,00 €
    1 * 1 ST
       14,00 €
  • 705-0126
    Sample cuvettes, with lid, 10/20/25 ml marked
    Sample cuvettes, with lid, 10/20/25 ml marked
    25 ml
    1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 102,00 €
    102,00 €
    1 * 6 ST
       102,00 €