Cryogenic vial boxes, Cryule®, WHEATON®
These cryoboxes are made of high quality chip-board and spray coated with polyethylene.
- Stores 1 and 2 ml cryogenic vials
- Holds 81 freestanding or round bottom vials
Suitable for use with vapor phase of liquid nitrogen.
Frequently Bought Together