Vacuum filtration complete units, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
Filtration assemblies with tapered joints, metal clamp and support base.
- Recommended for general filtration
- Units have a hose connection for ¼" (6 mm) Int.Ø flexible tubing
Recommended for routine filtration of organic solvents, corrosive liquids and the removal of particulates from HPLC solvents. Ground glass connection eliminates phthalate contamination that can occur from silicone or neoprene stoppers. Drip tip on the support bases extends below the hose connection preventing filtrate from entering the vacuum line.
Glass components manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirement.
Delivery information: Includes a coarse porosity (40 to 60 µm) fritted glass support base, graduated funnel, anodized aluminum clamp and 1 litre filter flask.
Frequently Bought Together