Precision probes for 735
Supplier: Testo
Jump to:
About this item
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- RangeAccuracyØ×LAvailabilityPrice
- 628-3299TC type T flexible air probe with PTFE cableTC type T flexible air probe with PTFE cable−50…+250 °C±0,3 °C (−20...+70 °C)
±(0,7 °C (±0,5% mv)1,5×2000 mmLog in for availability1 * 1 ST50,00 € - 620-3325PT100 glass-coated laboratory probe with replaceable glass pipePT100 glass-coated laboratory probe with replaceable glass pipe−50…+400 °C±0,2 °C
±0,2% mv (+200...+800 °C)6×200 mmLog in for availability1 * 1 ST134,00 € - 620-0216Highly accurate immersion PT100 probe with certificateHighly accurate immersion PT100 probe with certificate−40…+300 °C±0,3 °C (−80... −40 °C) ;
±0,1 °C + 0,05% mv (−40...0 °C)
±0,05 °C (0 ...100 °C)
±0,05 °C + 0,05 % mv (+100...+300 °C)4×295 mmLog in for availability1 * 1 ST472,00 €