Orders LinkContactUsLinkComponent
Thermometer probes
Anbieter:   COMARK
Thermometer probes
Anbieter:  COMARK
Certificates
Thermometer probes
8 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Description
    Range
    Application
    Ø×L
    Verfügbarkeit
    Preis
  • Mineral insulated semi flexible type K probe, with sub-miniature connector, AK21M
    −100…+850 °C
    General purpose air
    1,5×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 310,00 €
    310,00 €
    1 * 1 ST
       310,00 €
  • Flexible wire type K air probe, with sub-miniature connector, AK28M
    −100…+250 °C
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,90 €
    76,90 €
    1 * 1 ST
       76,90 €
  • Flexible wire type K air probe, with sub-miniature connector, AK29M
    −100…+250 °C
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,50 €
    80,50 €
    1 * 1 ST
       80,50 €
  • Flexible wire type K air probe, with sub-miniature connector, AK31M
    −100…+250 °C
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,50 €
    68,50 €
    1 * 1 ST
       68,50 €
  • Type T high temperature air probe up to +400 °C, sub-miniature connector, AK33M
    −100…+400 °C
    - mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,90 €
    66,90 €
    1 * 1 ST
       66,90 €
  • Type K industrial probe, with sub-miniature connector, IK21M
    −100...+850 °C
    Immersion
    1,5×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 272,00 €
    272,00 €
    1 * 1 ST
       272,00 €
  • Fast response type K probe, sub-miniature connector, PK24M
    −50…+250 °C
    Penetration
    3,3×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 124,00 €
    124,00 €
    1 * 1 ST
       124,00 €
  • Fast response type K probe for up to +250 °C, subminiature connector, SK21M
    −50...+250 °C
    Surface
    2,5×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 427,00 €
    427,00 €
    1 * 1 ST
       427,00 €