Orders LinkContactUsLinkComponent
Counting chambers
Counting chambers
Counting chambers
Supplier:  MARIENFELD
Certificates
Counting chambers
Counting chambers
3 Options Available Below
Product Details & Documents
These counting chambers are suitable for counting leucocytes and erythrocytes.
  • Double net ruling
  • Two counting grids with one division
  • Different types of design available
Certifications: IVD 98/79/EC.
Delivery information: Supplied with two cover glasses of 0,4 mm thickness.
Documents
Product Family Options
Product Information
  • Type
    Depth of chamber
    Availability
    Price
  • 630-1510
    Counting chamber, dark lined
    Counting chamber, dark lined
    Fuchs-Rosenthal
    0,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,73 €
    36,73 €
    1 * 1 ST
       36,73 €
  • 630-1511
    Counting chamber, dark lined
    Counting chamber, dark lined
    Nageotte
    0,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,40 €
    112,40 €
    1 * 1 ST
       112,40 €
  • 630-1509
    Counting chamber, bright lined
    Counting chamber, bright lined
    Imp. Neubauer
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,76 €
    73,76 €
    1 * 1 ST
       73,76 €