ContactUsLinkComponent
VWR®, Slide Mailers
VWR®, Slide Mailers
  631-1515
undefined
VWR®, Slide Mailers
  631-1515
  • No. of slides:
    5
  • Colour:
    Natural
  • Description:
    Slide mailers, with side opening
  • Material:
    PP
  • Seq:
    02
  • W×D×H:
    81×19×31 mm
  • Pk:
    25
These boxes are designed for shipping, storing, or staining slides.
  • Inside wall features slotted channels to keep slides separated and making removal easier
  • Closes securely to keep slides safe - side opening
  • Assorted colours (red, yellow, blue, purple, orange) for easier identification