Racks for orbital shakers SM 30 A, SM 30 A control, SM 30 B and SM 30 B control
Supplier: Edmund Bühler GmbH
Certificates
Racks for orbital shakers SM 30 A, SM 30 A control, SM 30 B and SM 30 B control
Supplier: Edmund Bühler GmbH
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for hotplates, VHP-C Series
Supplier: IKA
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Electronic contact thermometers, ETS-D5/ ETS-D6
Supplier: IKA
Cleanroom face masks, Kimtech™ M3, PE
Supplier: KIMBERLY CLARK
Magnetic stirrers, MICROSTIRRER
Supplier: VELP SCIENTIFIC
VWR®, Disposable Pestles, Microtubes and Cordless Motor for Pellet Mixing
Supplier: VWR Collection
Vortex mixers, MS 3
Supplier: IKA
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Disposable weighing dishes
Supplier: LUDI
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
VWR®, Vortex Mixer
Supplier: VWR Collection
Accessories for tube mill control
Supplier: IKA
Orbital/reciprocating shakers, SM 30 A and SM 30 A control / SM 30 C and SM 30 C control
Supplier: Edmund Bühler GmbH
VWR®, Magnetic Stirrer, Lab Disc
Supplier: VWR Collection
Combination pH electrodes, sensION™+
Supplier: Hach
Masterflex® Direct-Read Variable-Area Flowmeters with Valve, Metric Units, Avantor®
Supplier: Avantor Fluid Handling