Order Entry
Austria
Orders LinkContactUsLinkComponent
DNA prep for PCR - tissue, Extracta™

DNA prep for PCR - tissue, Extracta™

DNA prep for PCR - tissue, Extracta™
Supplier:  Quantabio
Certificates

3 Options Available Below

Product Details & Documents

The Extracta™ DNA prep for PCR - tissue consists of a two component reagent kit for rapid and efficient extraction of PCR-ready genomic DNA from mammalian tissues. The kit has been demonstrated to work with mouse tail biopsies, ear punches and a variety of human tissue samples including hair, buccal cells and saliva.

  • Simple, fast protocol
  • Compatible with laboratory automation
  • DNA is suitable for qPCR, high resolution melt analysis, endpoint PCR and other applications
  • Works with wide range of tissue
  • Optional 10 minute protocol for high throughput applications

The genomic DNA is suitable for sensitive downstream PCR applications including end-point PCR, High Resolution Melt Analysis (HRM) and quantitative real time PCR (qPCR) without requiring any additional clean-up. In addition, the extracted DNA may be used in multiplexed PCR applications such as transgene or knock-out analyses. Tissue extractions can be performed in tubes, plates or deep well blocks to allow for adaptation to workflow and automation on liquid handling workstations.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 733-2212
  • 733-2160
  • QUNT95091-002
  • Description:
  • Extracta™ DNA prep for PCR - tissue
  • Extracta™ DNA prep for PCR - tissue
  • Extracta™ DNA prep for PCR - tissue, 2 ml
  • Target molecule:
  • DNA
  • DNA
  • DNA
  • Refine target molecule:
  • Total DNA
  • Total DNA
  • Total DNA
  • Sample type:
  • Cultured cells|Human/animal cells and tissues
  • Cultured cells|Human/animal cells and tissues
  • Cultured cells|Human/animal cells and tissues
  • Method / Format:
  • Extraction for direct PCR
  • Extraction for direct PCR
  • Extraction for direct PCR
  • Prep size:
  • Scalable
  • Scalable
  • Scalable

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 733-2212
    Extracta™ DNA prep for PCR - tissue
    Extracta™ DNA prep for PCR - tissue
    250 mL
    Log in for availability
    Log in for availability
    - 1 330,00 €
    1 330,00 €
       1 330,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    733-2212
    Description
    Extracta™ DNA prep for PCR - tissue
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Cultured cells|Human/animal cells and tissues
    Method / Format
    Extraction for direct PCR
    Prep size
    Scalable
  • 733-2160
    Extracta™ DNA prep for PCR - tissue
    Extracta™ DNA prep for PCR - tissue
    25 mL
    Log in for availability
    Log in for availability
    - 158,00 €
    158,00 €
       158,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    733-2160
    Description
    Extracta™ DNA prep for PCR - tissue
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Cultured cells|Human/animal cells and tissues
    Method / Format
    Extraction for direct PCR
    Prep size
    Scalable
  • QUNT95091-002
    Extracta™ DNA prep for PCR - tissue, 2 ml
    Extracta™ DNA prep for PCR - tissue, 2 ml
    1
    Log in for availability
    Log in for availability
    - 16,50 €
    16,50 €
       16,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    QUNT95091-002
    Description
    Extracta™ DNA prep for PCR - tissue, 2 ml
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Cultured cells|Human/animal cells and tissues
    Method / Format
    Extraction for direct PCR
    Prep size
    Scalable
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.