Calibration weight sets, OIML class E2
Supplier: Sartorius Balances
Jump to:
About this item
These E2 class weight sets from 1 mg to 10 kg are made of non-magnetisable, stainless steel.
- Density of 8,0 g/dm³
- Knob weights highly polished
- Highly corrosion-resistant
: Up to 20 g supplied in a plastic case; from 50 g and above in a wooden case; from 1 kg and above, additionally supplied with gloves; Service weight set in a plastic carrying case.
16 Options Available Below
Product Details & Documents
These E2 class weight sets from 1 mg to 10 kg are made of non-magnetisable, stainless steel.
- Density of 8,0 g/dm³
- Knob weights highly polished
- Highly corrosion-resistant
: Up to 20 g supplied in a plastic case; from 50 g and above in a wooden case; from 1 kg and above, additionally supplied with gloves; Service weight set in a plastic carrying case.
Product Family Options
E2 Set of weights without certificates
Product Information
- Weight setTypeAvailabilityPrice
- 0,001 − 1000 gWire weight set, with gloves, without certificateLog in for availability1 * 1 ST4 000,00 €
- 0,001 − 5000 gWire weight set, with gloves, without certificateLog in for availability1 * 1 ST7 430,00 €
- 1 − 1000 gKnob weight set with gloves, without certificateLog in for availability1 * 1 SET3 210,00 €
- 1 − 5000 gKnob weight set with gloves, without certificateLog in for availability1 * 1 SET6 770,00 €
- 1 − 10000 gKnob weight set with gloves, without certificateLog in for availability1 * 1 SET8 230,00 €
E2 Set of weights with DKD certificates
Product Information
- Weight setTypeAvailabilityPrice
- 0,001 − 1000 gWire weight set, with gloves and DKD certificateLog in for availability1 * 1 ST4 740,00 €
- 0,001 − 5000 gWire weight set with gloves and DKD certificateLog in for availability1 * 1 ST8 420,00 €
- 1 − 1000 gKnob weight set with gloves and DKD certificateLog in for availability1 * 1 SET3 650,00 €
- 1 − 5000 gKnob weight set with gloves and DKD certificateLog in for availability1 * 1 SET7 410,00 €
- 1 − 10000 gKnob weight set with gloves and DKD certificateLog in for availability1 * 1 SET8 890,00 €