Orders LinkContactUsLinkComponent
Calibration weight sets, OIML class E1
Calibration weight sets, OIML class E1
Supplier:   Sartorius Balances
Calibration weight sets, OIML class E1
Supplier:  Sartorius Balances
Certificates
Calibration weight sets, OIML class E1
Calibration weight sets, OIML class E1
16 Options Available Below
Product Details & Documents
These E1 class weight sets from 1 mg to 10 kg are made of non-magnetisable, stainless steel.
  • Density of 8,0 g/dm³
  • Knob weights highly polished
  • Highly corrosion-resistant
Delivery information: Up to 20 g supplied in a plastic case; from 50 g and up in a wooden case; from 1 kg and up, additionally supplied with gloves.
Product Family Options
E1 set of weights in wooden boxes
Product Information
  • Weight set
    Type
    Availability
    Price
  • 0,001 − 5 g
    Wire weight set without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 990,00 €
    2 990,00 €
    1 * 1 ST
       2 990,00 €
  • 0,001 − 100 g
    Wire weight set without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 310,00 €
    4 310,00 €
    1 * 1 ST
       4 310,00 €
  • 0,001 − 200 g
    Wire weight set without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 290,00 €
    5 290,00 €
    1 * 1 ST
       5 290,00 €
  • 0,001 − 1000 g
    Wire weight set, with gloves, without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 140,00 €
    7 140,00 €
    1 * 1 ST
       7 140,00 €
  • 0,001 − 5000 g
    Wire weight set, with gloves, without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13 620,00 €
    13 620,00 €
    1 * 1 ST
       13 620,00 €
  • 1 − 1000 g
    Knob weight set with gloves, without certificate
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 5 570,00 €
    5 570,00 €
    1 * 1 SET
       5 570,00 €
  • 1 − 5000 g
    Knob weight set with gloves, without certificate
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 11 120,00 €
    11 120,00 €
    1 * 1 SET
       11 120,00 €
  • 1 − 10000 g
    Knob weight set with gloves, without certificate
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 14 960,00 €
    14 960,00 €
    1 * 1 SET
       14 960,00 €
E1 set of weights in wooden boxes with DKD certificates
Product Information
  • Weight set
    Type
    Availability
    Price
  • 0,001 − 5 g
    Wire weight set with DKD certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 280,00 €
    4 280,00 €
    1 * 1 ST
       4 280,00 €
  • 0,001 − 100 g
    Wire weight set with DKD certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 260,00 €
    6 260,00 €
    1 * 1 ST
       6 260,00 €
  • 0,001 − 200 g
    Wire weight set with DKD certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 400,00 €
    7 400,00 €
    1 * 1 ST
       7 400,00 €
  • 0,001 − 1000 g
    Wire weight set, with gloves and DKD certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9 430,00 €
    9 430,00 €
    1 * 1 ST
       9 430,00 €
  • 0,001 − 5000 g
    Wire weight set with gloves and DKD certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16 750,00 €
    16 750,00 €
    1 * 1 ST
       16 750,00 €
  • 1 − 1000 g
    Knob weight set with gloves and DKD certificate
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 6 930,00 €
    6 930,00 €
    1 * 1 SET
       6 930,00 €
  • 1 − 5000 g
    Knob weight set with gloves and DKD certificate
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 13 150,00 €
    13 150,00 €
    1 * 1 SET
       13 150,00 €
  • 1 − 10000 g
    Knob weight set with gloves and DKD certificate
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 17 250,00 €
    17 250,00 €
    1 * 1 SET
       17 250,00 €