Order Entry
Austria
ContactUsLinkComponent
Staining jars according to Coplin, WHEATON®
Staining jars according to Coplin, WHEATON®
  631-2508
undefined
Staining jars according to Coplin, WHEATON®
  631-2508

 

  • Description:
    Staining jar acc. to Coplin, with glass lid
  • W×D×H:
    26×26×90 mm
  • Pk:
    6

 

 

Soda-lime glass.

  • Heavy glass walls and broad base for increased stability
  • Requires low reagent volume
  • Capacity: Approx. 65 ml

Staining jar for 5 single standard slides 76×26 mm vertically or 10 slides back to back with glass cover.