ContactUsLinkComponent
Sample bottles, narrrow neck, with screw cap, WHEATON®
Sample bottles, narrrow neck, with screw cap, WHEATON®
Catalog # WHEA990477
Supplier:  DWK Life Sciences
undefined
Sample bottles, narrrow neck, with screw cap, WHEATON®
Catalog # WHEA990477
Supplier:  DWK Life Sciences
Specifications
  • Thread:
    38-430
  • Capacity:
    1000 ml
  • Pk:
    1
Specifications
About this item
Glass, clear.
  • Excellent for accessing fluid flow in deep manholes and hard to reach sub-surface sampling areas
  • Helps meet EPA requirements for water and waste water sampling
  • 38-430 thread
Each sampler consists of an inert polypropylene head with stainless steel fittings and an aluminum pole assembly with a golden alodine finish. It comes with a 1000 mL narrow-mouth sample bottle with a PTFE- lined cap. To use it, screw a sample bottle into the head and lower to the desired depth. Pull the cable at the top of the handle which will lift a spring-loaded plunger from the bottle opening, allowing liquid to enter through four 5/8" diameter holes. Release the cable when the bottle is full to reseat the plunger.