Order Entry
Austria
ContactUsLinkComponent
 
Screw caps for sensors, BOLA
  BOHLD780-14
 :  Bohlender
undefined
Screw caps for sensors, BOLA
  BOHLD780-14
 :  Bohlender

 

  • Description:
    for inserting sensors Ø 12 mm (±0,5)
  • Pk:
    1

 

 

PPS/PTFE.

  • Temperature resistance: from 0 to +110 °C
  • Universal chemical resistance
  • For contamination-free insertion of sensors into bottles with GL 45 thread

Black screw cap made of PPS with GL 45 thread, movable body made of PTFE with adjusting screw for fixing and sealing sensors. The product is only exposed to PTFE.