Vial racks, Chromacol™
Jump to:
About this item
These vial racks are chemically resistant and fairly robust for a safe working position on the lab bench and during transport.
- Ideal for space saving storage in fridges
- Temperature resistant from −90 to 121 °C (except foam)
0 Options Available Below
Product Details & Documents
These vial racks are chemically resistant and fairly robust for a safe working position on the lab bench and during transport.
- Ideal for space saving storage in fridges
- Temperature resistant from −90 to 121 °C (except foam)
Product Family Options