Order Entry
Austria
ContactUsLinkComponent
Microbiology liquid handling system, DOSE IT
Microbiology liquid handling system, DOSE IT
  432-0056
 :  Integra
undefined
Microbiology liquid handling system, DOSE IT
  432-0056
 :  Integra

 

  • Flow rate:
    0,6 - 5000 ml/min
  • Weight:
    3,5 kg
  • Description:
    DOSE IT, complete system
  • W×D×H:
    210×191×203 mm
  • Pk:
    1

 

 

The DOSE IT peristaltic pump makes dispensing of culture media, buffers and other solutions easy and efficient. In addition to the dispensing and pump mode, DOSE IT is able to create customised liquid handling protocols. Easy to follow instructions on the display allow users to set up individual protocols, consisting of consecutive application steps.

  • Large display and clear user guidance for fast set-up and use
  • CUSTOMIZE mode for easy creation of personalised protocols consisting of up to 20 consecutive steps based on settable parameters, such as volume, flow rate and repetitions
  • The flow rate can be set to ramp up or down to the target speed, avoiding, for example, foaming, splashing or unwanted turbulences during dispensing
  • Memory for 20 liquid handling protocols for conveniently switching between different applications
  • Compact and portable design for minimal space consumption and maximum user friendliness

With its very small footprint and light weight, the DOSE IT fits everywhere in the laboratory and can be moved easily – no need to waste valuable bench space or to have a pump in every laboratory.

 : Complete DOSE IT set includes:
1× silicone tubing, inner diameter 4 mm (length 2,5 m)
2× aspiration/dispensing tubes length 10 cm, stainless steel, one end dented
1× aspiration/dispensing tube length 35 cm, stainless steel, one end dented
1× tube collar
1× pipette tubing connector
1× mains adapter