ContactUsLinkComponent
Large insulating dewar flasks, type 131 - 135
Large insulating dewar flasks, type 131 - 135
Supplier:  KGW
Certificates
undefined
Large insulating dewar flasks, type 131 - 135
Supplier:  KGW
Specifications
  • Catalog No:
  • KGWA1265
  • KGWA1263
  • Model:
  • 134
  • 132
  • Capacity:
  • 28 L
  • 14 L
  • Daily evaporation:
  • 0,6 L
  • 0,4 L
  • Overall height:
  • 765 mm
  • 635 mm