WHEATON® E-Z Ex-Traction™ High Recovery Vials
Supplier: DWK Life Sciences
Certificates
About this item
Precision-manufactured from low extractable borosilicate glass, these vials are ideal for compound biological sample applications. Vials conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
- Conical well ensures maximum sample recovery
- Quality engineered for automated sample handling systems
Related Information
Screw caps
Product Information
Frequently Bought Together