ContactUsLinkComponent
VWR® Thermotech™, Beakers, Low Form, Heatable
  213-0272
 :
undefined
VWR® Thermotech™, Beakers, Low Form, Heatable
  213-0272
 :
Select an Option
75 mm
Select an Option
250 ml
Select an Option
94 mm
Select an Option
02
Select an Option
1
PTFE, white with black PTFE carbon base.
  • Totally inert pure PTFE inner surface
  • Heatable to 270 °C without distortion
  • Improved heat transfer
Features the unique combination of a pure PTFE body with a specially formulated stabilised PTFE carbon base as a single moulding.
Frequently Bought Together