Order Entry
Austria
ContactUsLinkComponent
 
Desalting columns with Sephadex G-25 resin, HiTrap™
 :  Cytiva
 
undefined
Desalting columns with Sephadex G-25 resin, HiTrap™
 :  Cytiva
 

 

  • Catalog No:
  • 29-0486-84
  • 17-1408-01
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 2 - 13
  • 2 - 13
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

 

Frequently Bought Together