Affinity chromatography columns, HiTrap™ NHS-Activated HP
: Cytiva
HiTrap NHS-activated HP prepacked chromatography columns allow you to design an affinity column suitable for your purification needs.
- Easy coupling of any ligand containing primary amine groups
- Packed with NHS-Activated Sepharose High Performance resin
- Convenient prepacked columns can be used with a syringe, pump, or chromatographic system, such as ÄKTA systems
This NHS-activated resin can be used with any primary amine-containing ligand. The resin reacts with primary amine groups to form stable amide linkages that covalently immobilize antibodies or other proteins for affinity purification. The resin enables simple coupling, carried out at close to neutral pH.
The NHS-activated Sepharose High Performance (HP) resin in these columns is delivered in 100% isopropanol to protect the active groups. It contains ~ 10 μmol NHS groups/mL resin.
- Catalog No:
- 17-0716-01
- 17-0717-01
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 3 - 12
- 3 - 12
- Storage:
- 4 to 8 °C, 100% Isopropanol
- 4 to 8 °C, 100% Isopropanol
Frequently Bought Together