ContactUsLinkComponent
MagRack™ Maxi
MagRack™ Maxi
Catalog # 28-9864-41
Supplier:  Cytiva
undefined
MagRack™ Maxi
Catalog # 28-9864-41
Supplier:  Cytiva
Specifications
  • For tubes:
    1×50 ml + 1×15 ml
  • No. of Holes:
    2
  • Description:
    MagRack™ Maxi
  • Pk:
    1
Specifications
About this item
MagRack Maxi is a magnetic rack for small-scale protein purification and sample enrichment with magnetic beads used for sample volumes up to 50 ml.
  • Provides convenience by making it easy to perform small-scale purification and enrichment of sample volumes up to 50 ml
  • Allows for optimal performance during protein purifications or enrichments
  • Each rack consists of an aluminium housing (blue) and a detachable plastic bar (white) containing a neodymium magnet
The anodized aluminium housing assists with electrical conductivity whilst the neodymium magnet is extraordinarily strong, helping with magnetic reaction times as well as optimizing performance during protein purifications or enrichments.
MagRack Maxi, is complementary to the following magnetic bead-based products: Protein A Mag Sepharose, Protein A Mag Sepharose Xtra, Protein G Mag Sepharose, Protein G Mag Sepharose Xtra, His Mag Sepharose Ni, NHS Mag Sepharose, and Streptavidin Mag Sepharose.
MagRack Maxi and MagRack 6 cover a broad range of sample volumes from low-microliter to high-milliter purification scales, enhancing the flexibility of magnetic beads separations.