Order Entry
Austria
ContactUsLinkComponent
Dye terminator removal, Mag-Bind® SeqDTR™
Dye terminator removal, Mag-Bind® SeqDTR™
Supplier:  OMEGA BIO-TEK
Certificates
undefined
Dye terminator removal, Mag-Bind® SeqDTR™
Supplier:  OMEGA BIO-TEK
Certificates

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Sample size:
    Variable
  • Description:
    Mag-Bind™ SeqDTR™

Specifications

Related Information

Frequently Bought Together