ContactUsLinkComponent
Scrubbing sponge for glassware, Cleanware™, SP Bel-Art
Scrubbing sponge for glassware, Cleanware™, SP Bel-Art
Catalog # BELAF170780001
undefined
Scrubbing sponge for glassware, Cleanware™, SP Bel-Art
Catalog # BELAF170780001
Specifications
  • Packed:
    2 sponges/pack
  • Colour:
    Yellow/green
  • L×W:
    108×64 mm
  • Thickness:
    25 mm
  • Pk:
    2
Specifications
About this item
This odor-resistant, highly absorbent sponge is designed to remove stubborn residue on glassware.
  • Wipe dry performance equal to cellulose; maintains the desirable physical properties of polyurethane
  • Foam contains anti-microbial technology to inhibit odor causing bacteria within the sponge itself
  • Stays soft and flexible, wet or dry; will not crack or shrink, or become brittle like cellulose
  • Scouring side resists snagging; does not leave fibers behind and retains aggressive scouring ability even with frequent use