Orders LinkContactUsLinkComponent
VWR®, Calibration Weight Sets, Class E2
Supplier:   VWR Collection
VWR®, Calibration Weight Sets, Class E2
Supplier:  VWR Collection
Certificates
VWR®, Calibration Weight Sets, Class E2
VWR®, Calibration Weight Sets, Class E2
41 Options Available Below
Product Details & Documents
Weights made from solid, anti-magnetic, polished stainless steel. Suitable for high resolution electronic balances.
  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate
Conform to OILM R111 recommendations.
 : Conform to OILM R111 recommendations.
 : All weight sets listed in the 'set contents' table are available with or without COFRAC certificate - please visit website or contact your local Avantor sales office. Weights are also available individually - please search for product 'Calibration weights, Class E2'.
Product Family Options
E2 weight sets in wooden boxes with certificate
Product Information
  • Weight set
    No. of weights
    Total weight
    Availability
    Price
  • 1 mg - 500 mg
    12
    1,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
  • 1 mg - 50 g
    20
    111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 320,00 €
    2 320,00 €
    1 * 1 ST
       2 320,00 €
  • 1 mg - 100 g
    21
    211,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 750,00 €
    2 750,00 €
    1 * 1 ST
       2 750,00 €
  • 1 mg - 200 g
    23
    611,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 150,00 €
    3 150,00 €
    1 * 1 ST
       3 150,00 €
  • 1 mg - 500 g
    24
    1111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 300,00 €
    3 300,00 €
    1 * 1 ST
       3 300,00 €
  • 1 mg - 1 kg
    25
    2111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 930,00 €
    3 930,00 €
    1 * 1 ST
       3 930,00 €
  • 1 mg - 2 kg
    27
    6111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 130,00 €
    5 130,00 €
    1 * 1 ST
       5 130,00 €
  • 1 mg - 5 kg
    28
    11111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 250,00 €
    6 250,00 €
    1 * 1 ST
       6 250,00 €
  • 1 g - 50 g
    8
    110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 260,00 €
    1 260,00 €
    1 * 1 ST
       1 260,00 €
  • 1 g - 100 g
    9
    210 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 450,00 €
    1 450,00 €
    1 * 1 ST
       1 450,00 €
  • 1 g - 200 g
    11
    610 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 910,00 €
    1 910,00 €
    1 * 1 ST
       1 910,00 €
  • 1 g - 500 g
    12
    1110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 380,00 €
    2 380,00 €
    1 * 1 ST
       2 380,00 €
  • 1 g - 1 kg
    13
    2110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 770,00 €
    2 770,00 €
    1 * 1 ST
       2 770,00 €
  • 1 g - 2 kg
    15
    6110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 260,00 €
    4 260,00 €
    1 * 1 ST
       4 260,00 €
  • 1 g - 5 kg
    16
    11110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 390,00 €
    5 390,00 €
    1 * 1 ST
       5 390,00 €
E2 weight sets in plastic cases with certificate
Product Information
  • Weight set
    No. of weights
    Total weight
    Availability
    Price
  • 1 g - 10 kg
    17
    21110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 900,00 €
    6 900,00 €
    1 * 1 ST
       6 900,00 €
  • 1 mg - 10 kg
    29
    21111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 700,00 €
    7 700,00 €
    1 * 1 ST
       7 700,00 €
  • 1 mg - 500 mg
    12
    1,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 849,00 €
    849,00 €
    1 * 1 ST
       849,00 €
  • 1 mg - 100 g
    21
    211,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 270,00 €
    2 270,00 €
    1 * 1 ST
       2 270,00 €
  • 1 mg - 200 g
    23
    611,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 570,00 €
    2 570,00 €
    1 * 1 ST
       2 570,00 €
  • 1 mg - 500 g
    24
    1111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 800,00 €
    2 800,00 €
    1 * 1 ST
       2 800,00 €
  • 1 g - 100 g
    9
    210 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 350,00 €
    1 350,00 €
    1 * 1 ST
       1 350,00 €
  • 1 g - 200 g
    11
    610 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 810,00 €
    1 810,00 €
    1 * 1 ST
       1 810,00 €
  • 1 g - 500 g
    12
    1110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 150,00 €
    2 150,00 €
    1 * 1 ST
       2 150,00 €
E2 weight sets in wooden boxes without certificate
Product Information
  • Weight set
    No. of weights
    Total weight
    Availability
    Price
  • 1 mg - 500 mg
    12
    1,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 577,00 €
    577,00 €
    1 * 1 ST
       577,00 €
  • 1 mg - 50 g
    20
    111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 500,00 €
    1 500,00 €
    1 * 1 ST
       1 500,00 €
  • 1 mg - 100 g
    21
    211,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 840,00 €
    1 840,00 €
    1 * 1 ST
       1 840,00 €
  • 1 mg - 200 g
    23
    611,11
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 330,00 €
    2 330,00 €
    1 * 1 ST
       2 330,00 €
  • 1 mg - 500 g
    24
    1111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 660,00 €
    2 660,00 €
    1 * 1 ST
       2 660,00 €
  • 1 mg - 1 kg
    25
    2111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 000,00 €
    3 000,00 €
    1 * 1 ST
       3 000,00 €
  • 1 mg - 2 kg
    27
    6111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 040,00 €
    4 040,00 €
    1 * 1 ST
       4 040,00 €
  • 1 mg - 5 kg
    28
    11111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 220,00 €
    5 220,00 €
    1 * 1 ST
       5 220,00 €
  • 1 g - 50 g
    8
    110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 951,00 €
    951,00 €
    1 * 1 ST
       951,00 €
  • 1 g - 100 g
    9
    210 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 080,00 €
    1 080,00 €
    1 * 1 ST
       1 080,00 €
  • 1 g - 200 g
    11
    610 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 460,00 €
    1 460,00 €
    1 * 1 ST
       1 460,00 €
  • 1 g - 500 g
    12
    1110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 900,00 €
    1 900,00 €
    1 * 1 ST
       1 900,00 €
  • 1 g - 1 kg
    13
    2110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 270,00 €
    2 270,00 €
    1 * 1 ST
       2 270,00 €
  • 1 g - 2 kg
    15
    6110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 660,00 €
    3 660,00 €
    1 * 1 ST
       3 660,00 €
  • 1 g - 5 kg
    16
    11110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 700,00 €
    4 700,00 €
    1 * 1 ST
       4 700,00 €
Weight Sets in Plastic Cases without Certificate
Product Information
  • Weight set
    No. of weights
    Total weight
    Availability
    Price
  • 1 g - 10 kg
    17
    21110 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 270,00 €
    6 270,00 €
    1 * 1 ST
       6 270,00 €
  • 1 mg - 10 kg
    29
    21111,11 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 680,00 €
    6 680,00 €
    1 * 1 ST
       6 680,00 €