Double spatulas, micro, with flat ends, SP Bel-Art
Stainless steel coated with Fluo-Kem® Teflon® FEP.
- With flat ends (7,9×50 mm)
- Complies with FDA regulations governing direct food contact applications
- Available with straight or tapered blade
Frequently Bought Together